EMD Millipore Durapore™ PVDF Membrane Filters: Hydrophilic: 0.65μ Pore Size
Recommended when maximum recovery is important
Manufacturer:  EMD Millipore DVPP09050
 View more versions of this product
Catalog No. DVPP09050
Specifications Description & Specifications
Specifications
Polyvinylidene Fluoride (PVDF)
White
Yes
69.0mL/min./cm sq.
90mm
50/pk.
14psi
Hydrophilic
Accessories