EMD Millipore Durapore™ PVDF Membrane Filters: Hydrophilic: 0.10μ Pore Size
Recommended when maximum recovery is important
Manufacturer:  EMD Millipore VVLP01300
 View more versions of this product
Catalog No. VVLP01300
Specifications Description & Specifications
Specifications
Polyvinylidene Fluoride (PVDF)
White
Yes
2.5mL/min./cm sq.
13mm
100/pk.
70psi
Hydrophilic