MilliporeSigma™ Durapore™ Membrane Filter
Strength, flexibility, and broad chemical compatibility for critical biological separations
$384.00 - $569.00
Specifications
Color White
View More Specs
Products 3
Catalog Number Mfr. No. Diameter (Metric) Outside For Use With (Application) Material Pore Size Price Quantity  
Catalog Number Mfr. No. Diameter (Metric) Outside For Use With (Application) Material Pore Size Price Quantity  
GVWP2932A
View Documents
MilliporeSigma™
GVWP2932A
29.3cm Provide strength, flexibility, and broad chemical compatibility for critical biological separations PVDF 0.22μm
Pack of 25 Each for $550.00
HVLP2932A
View Documents
MilliporeSigma™
HVLP2932A
29.3cm Provide strength, flexibility, and broad chemical compatibility for critical biological separations PVDF 0.45μm
Pack of 25 Each for $569.00
SVLP09050
View Documents
MilliporeSigma™
SVLP09050
90mm For clarifying filtration of biological solution; particle monitoring Polyvinylidene Fluoride (PVDF) 5.0μm
Each for $384.00
Description
Description
  • Protein binding to hydrophilic Durapore membranes is two orders of magnitude lower than nitro-cellulose or PTFE
Specifications
Specifications
White
SDS