Please login to your online account to display your discounted pricing

EMD Millipore™ Dulbecco's Modified Eagle Media 1X, Liquid, w/4,500mg/L Glucose


Manufacturer: emd millipore corporation  SLM020A

 View more versions of this product

Catalog No. SLM020A

  • / Each
 

Description & Specifications

Specifications

Quantity 1L
Storage Requirements 2-8°C for 10 months
For Use With (Application) Cell culture