EMD Millipore™ Dulbecco's Modified Eagle Media 1X, Liquid, w/4,500mg/L Glucose
Manufacturer:  Emd Millipore Corporation SLM020B
 View more versions of this product
Catalog No. SLM020B
Specifications Description & Specifications
Specifications
Cell culture
500mL
2-8°C for 10 months