Please login to your online account to display your discounted pricing

EMD Millipore Direct-Q™ 5 Tap to Pure and Ultrapure Water Purification System

Provide Type I and Type III water from potable tap water with the Direct-Q™ 5 Tap to Pure and Ultrapure Water Purification System.

$6,282.00 - $7,374.00

Specifications

Bacterial retention Greater than99%
Contaminant Rejection (Microbiological) Greater than99%
Contaminant Rejection (Organic) Greater than99%
Contaminant Rejection (Particulate) Greater than96%
Bacterial Content Less than0.1cfu/mL
View More Specs
Products
Catalog Number Mfr. No. Certifications/Compliance Model Product Type Product Water Type Price Quantity    

ZRQSVP500

 
emd millipore
ZRQSVP500
UL, cUL, CE, ISO 9001, ISO 14001 Direct-Q™ 5 Pure water purification system Type 3 Each for $6,282.00

ZRQSVR500

 
emd millipore
ZRQSVR500
UL, cUL, CE, ISO 9001, ISO 14001 Direct-Q™ 5UV Pure water purification system Type 3 Each for $7,374.00

ZRQSVR5US

 
emd millipore
ZRQSVR5US
UL, cUL Direct-Q™ 5UV Ultrapure water purification system Type 1 Each for $7,374.00

ZRQSVP5US

 
emd millipore
ZRQSVP5US
UL, cUL Direct-Q™ 5 Ultrapure water purification system Type 1 Each for $6,282.00
Description & Specifications

Specifications

Bacterial retention Greater than99%
Contaminant Rejection (Microbiological) Greater than99%
Contaminant Rejection (Organic) Greater than99%
Contaminant Rejection (Particulate) Greater than96%
Bacterial Content Less than0.1cfu/mL
Daily Output 1 to 10L
Endotoxin Concentration Less than0.001
Feedwater Source Reverse osmosis tap water
Flow Rate 5
Electrical Requirements 100/240V 50/60Hz
For Use With (Application) HPLC, GC, ILC and TOC analyses
Hertz 50/60Hz
Particle Size Greater than0.22μm, Less than1 Particulate/mL
Voltage 100/240V

Ideal for laboratories that do not have easy access to pretreated water and need between 1 and 10 L of ultrapure water per day.

  • Produce Type 1 ultrapure water (18.2 Mωcm resistivity at 25°C and >10 ppb Total Organic Carbon) on demand directly from potable tap water
  • Available with a built-in 185 and 254nm UV lamp to produce low TOC water required by organic-sensitive applications
  • Produce Type 3 (Reverse Osmosis) water for basic lab applications at a flow rate of 3, 5, or 8L/hr at 15°C
  • Type 3 water is easily available from external 30/60 L reservoirs
  • Maintenance reduced to a single cartridge change once or twice a year
  • Alarm warning in case of a sudden drop in the quality of the product water
  • Dispensing adapted to different size laboratory glassware
  • Time-saving fixed volume dispense

  • Type 1 water for HPLC mobile phase preparation, sample dilution, buffer and cell culture media preparation, preparation of chemical solutions used with titrators, spectrophotometers and electrophoresis systems, preparation of culture media and reagents for molecular biology applications
  • Type 3 water for non-critical applications such as glassware washing or rinsing

UL listed

cUL listed