Please login to your online account to display your discounted pricing

EMD Millipore Direct-Q™ 5 Tap to Pure and Ultrapure Water Purification System


Provide Type I and Type III water from potable tap water with the Direct-Q™ 5 Tap to Pure and Ultrapure Water Purification System.

Manufacturer: emd millipore  ZRQSVP5US

 View more versions of this product

Catalog No. ZRQSVP5US

  • / Each
 

Description & Specifications

Specifications

Particle Size Greater than0.22μm, Less than1 Particulate/mL
Product Type Ultrapure water purification system
Bacterial retention Greater than99%
Hertz 50/60Hz
Flow Rate 5
Bacterial Content Less than0.1cfu/mL
Contaminant Rejection (Organic) Greater than99%
Endotoxin Concentration Less than0.001
Feedwater Source Reverse osmosis tap water
Electrical Requirements 100/240V 50/60Hz
Contaminant Rejection (Microbiological) Greater than99%
For Use With (Application) HPLC, GC, ILC and TOC analyses
Certifications/Compliance UL, cUL
Voltage 100/240V
Model Direct-Q™ 5
Daily Output 1 to 10L
Product Water Type Type 1
Contaminant Rejection (Particulate) Greater than96%