Please login to your online account to display your discounted pricing

EMD Millipore™ Cytochrome c, Equine Heart, Protein

$144.00 - $144.00

Specifications

Immunogen Cytochrome c
Format Salt-free lyophilized solid
Storage Requirements Following reconstitution, refrigerate (4deg;C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°C, for up to 6 months at −20°C, and for up to 3 days at room temperature.
Regulatory Status RUO
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

25-060-0100MG

 
emd millipore corporation
250600100MG
100mg Each for $144.00
Description & Specifications

Specifications

Immunogen Cytochrome c
Format Salt-free lyophilized solid
Storage Requirements Following reconstitution, refrigerate (4deg;C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 2 weeks at 4°C, for up to 6 months at −20°C, and for up to 3 days at room temperature.
Regulatory Status RUO

Native cytochrome c purified from equine heart. Peripheral protein found on outer face of inner mitochondrial membrane. Useful as a mitochondrial marker. Plays an important role in electron transport.