MilliporeSigma™ Chromolith™ SemiPrep Guard Cartridges
For prolonging the lifespan of Chromolith™ analytical columns
Supplier: MilliporeSigma™ 1520360001
Description
Although the Chromolith™ columns are well known for their robustness and longevity, guard columns are available to prolong the lifespan of Chromolith™ analytical columns even further. Package includes three guard cartridges.Specifications
1/Pk. | |
HPLC Guard Cartridge | |
SemiPrep RP-18 endcapped sorbent |
10 mm | |
10 mm |