MilliporeSigma™ Chromolith™ RP Endcapped HPLC Guard Cartridges
Also popular for the protection of classical packed HPLC columns
Supplier:  MilliporeSigma™ 1520050001
 View more versions of this product
Catalog No. M1520050001
Description
Description
  • Handles easily
  • Adds to the robustness and longevity of monolithic analytical columns
Specifications
Specifications
3/Pk.
HPLC Guard Cartridge
RP-18 endcapped sorbent
5 mm
3 mm