Please login to your online account to display your discounted pricing
EMD Millipore Chromolith™ RP Endcapped HPLC Guard Cartridge Starter Kits
Includes the holder and three guard cartridges
Manufacturer: emd millipore  1514710001
 View more versions of this product
Catalog No. M1514700001
  • / Each
Description & Specifications
Specifications
Product Type HPLC Guard Cartridge Kit
Sorbent RP-18 endcapped sorbent
Quantity 1/Pk.
Length (Metric) 5mm
Diameter (English) Inside 4.6mm