MilliporeSigma™ Chemicon™ Caspase 9 Colorimetric Activity Assay Kit, LEHD
Provide a simple and convenient means for assaying the activity of caspases that recognize the LEHD
Supplier: MilliporeSigma™ APT139
Specifications
-20°C | |
Colorimetric | |
pNA Standard, 5X Cell Lysis Buffer, 5X Assay Buffer, Caspase-9 Substrate, Caspase-9 Inhibitor |
Caspase 9 Colorimetric Activity Assay Kit, LEHD | |
Provide a simple and convenient means for assaying the activity of caspases that recognize the LEHD | |
25 Tests |