MilliporeSigma™ Certipur™ Refractive Index Standard Kits
Refractive index standards Certipur™ according to Ph Eur.
Manufacturer:  MilliporeSigma™ 1089630001
 View more versions of this product
Catalog No. M1089630001
Description
Description
These standards are for the calibration of refractometers. We offer different refractive index standards in order to perform calibration in the same range as the final measurement.
  • Stable solutions with minimum shelf life of 3 years
  • Ready-to-use solutions
  • Each standard kit is packed with a certificate of analysis
  • Applicable at 20°C and 25°C measuring temperature
  • According to Ph Eur. 2.2.6
Specifications
Specifications
1-Methyl-naphthalene/water, traceable to SRM from NIST and PTB n (20°C)=1,6160 Certipur