EMD Millipore Cell Culture Multiwell Plate
Enables most adherent cells to attach and proliferate
$234.14 - $234.14
Specifications
Sterile Yes
For Use With (Application) Cell attachment, cell culture, cell differentiation, immunocytochemistry
Length (Metric) Exterior 12.8cm
Material Polystyrene
Type Cell Culture Multiwell Plate
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. No. of Wells Price Quantity  
Catalog Number Mfr. No. No. of Wells Price Quantity  
PIMWS2450 EMD Millipore
PIMWS2450
24 Pack of 50 Each for $234.14
Specifications
Description
6-, 12- and 24-well formats provide users flexibility to run multiple samples simultaneously.
  • Can be used independently for general cell culture or in conjunction with Millicell™ inserts
  • Compatible with cellular and fluorescent staining processes
  • Can be easily prepared for SEM and TEM
  • Made of polystyrene
  • Sterile
Cell attachment, cell culture, cell differentiation, immunocytochemistry
Order Info
Individually blister packaged in packs of 50.
Description & Specifications
Specifications
Yes
12.8cm
Cell Culture Multiwell Plate
Cell attachment, cell culture, cell differentiation, immunocytochemistry
Polystyrene
8.55cm