Please login to your online account to display your discounted pricing

EMD Millipore™ Cell Culture Freezing Media with DMSO


Manufacturer: emd millipore corporation  S0025F

 View more versions of this product

Catalog No. S0025F

  • / Each
 

Description & Specifications

Specifications

Storage Requirements -20°C
For Use With (Application) Stem cell culture and cell culture
Quantity 50mL