MilliporeSigma™ Calbiochem™ Cell Culture Tested G 418 Sulfate
an aminoglycoside antibiotic related to Gentamicin
Manufacturer:  MilliporeSigma™ 345810250MG
 View more versions of this product
Catalog No. 345810250MG
Description
Description
G418 also known as Geneticin™ is an aminoglycoside antibiotic related to Gentamicin. Used as a selective agent in transfection of eukaryotic cells. Has highest potency ≥730μg/mg and purity ≥98%.
Specifications
Specifications
250mg
4 to 6
≥98% by TLC
15-30°C