Please login to your online account to display your discounted pricing

EMD Millipore Analytical Vacuum Filter Holders


Use to vacuum filter liquids for bacteriological or particulate contamination analysis

Manufacturer: emd millipore  XF2004725

 View more versions of this product

Catalog No. XF2004725

  • / Each
 

Description & Specifications

Specifications

Material Stainless Steel
Diameter (Metric) Inner 19.4cm
Height (Metric) 7.6cm
Capacity (Metric) 250mL
Surface Area (Metric) 9cm2