missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Ultra-2 Centrifugal Filter Unit

With 98% sample recovery

$151.00 - $151.00

Specifications

Diameter (Metric) 15.9mm
Membrane Material Regenerated Cellulose
For Use With (Application) For protein purification/extraction; western blotting
Surface Area 1cm2
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes MWCO Price Quantity  
Catalog Number Mfr. No. Includes MWCO Price Quantity  
UFC201024 EMD Millipore
UFC201024
With Ultracel-10 membrane; NMWL: 10kDa 10 kDa Pack of 24 Each for $151.00
UFC205024 EMD Millipore
UFC205024
With Ultracel-50 membrane; NMWL: 50kDa 50 kDa Pack of 24 Each for $151.00
UFC203024 EMD Millipore
UFC203024
With Ultracel-30 membrane; NMWL: 30kDa 30 kDa Pack of 24 Each for $151.00
UFC200324 EMD Millipore
UFC200324
With Ultracel-3 membrane; NMWL: 3kDa 3 kDa Each for $151.00
UFC210024 EMD Millipore
UFC210024
With Ultracel-100 membrane; NMWL: 100kDa 50 kDa Pack of 24 Each for $151.00
Specifications

Description

Provides a fast and easy way to concentrate, desalt and exchange the buffer for both proteins and nucleic acids. By incorporating familiar Amicon Ultra features such as the vertical membrane design and true dead stop, the Amicon Ultra 2mL filter delivers high recovery, short spin times and quality results even for the most dilute samples.

  • True engineered dead stop provides consistent recovery without spinning to dryness
  • Reverse spin aids in sample recovery, especially with small, dilute samples
  • Vertical membrane design aligns with the filtrate for less clogging and faster filtering
  • Ultracel low-binding membrane combines with the innovative vertical housing to provide fast sample processing without sacrificing recovery
  • Fast processing, with 10- to 30-minute spin times
Specifications
  • Concentrates 2mL down to 15-70μL
  • Filtration area: 1cm2
  • Diameter: 15.9mm
  • Length: 4.71 in. (119mm)
  • Color: clear
  • Filter type: ultrafiltration
  • Device material: styrene/butadiene (housing), polypropylene (collection tube)
  • Sold in pack of 24

Protein purification, Western blotting, Protein extraction

Description & Specifications

Specifications

15.9mm
Regenerated Cellulose
For protein purification/extraction; western blotting
1cm2