Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Ultra-2 Centrifugal Filter Unit


With 98% sample recovery

Manufacturer: emd millipore  UFC205024

 View more versions of this product

Catalog No. UFC205024

  • / Pack of 24 Each
 

Description & Specifications

Specifications

Volume (Metric) 2mL
Membrane Material Regenerated Cellulose
Diameter (Metric) 15.9mm
Surface Area 1cm2
Includes With Ultracel-50 membrane; NMWL: 50kDa
For Use With (Application) For protein purification/extraction; western blotting
MWCO 50 kDa