missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Amicon™ Ultra-2 Centrifugal Filter Units

Catalog No. UFC210024
Change view
Click to view available options
MWCO:
10 kDa
100 kDa
3 kDa
30 kDa
50 kDa
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. MWCO
UFC210024 100 kDa
UFC203024 30 kDa
UFC201024 10 kDa
UFC200324 3 kDa
UFC205024 50 kDa
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. UFC210024 Supplier MilliporeSigma™ Supplier No. UFC210024
Only null left

Includes

Two microcentrifuge tubes

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting.

Achieve both high sample recovery and fast sample processing. EMD Amicon™ Ultra Centrifugal Filter Units feature regenerated cellulose membranes that are ideal for protein & nucleic acid purification, concentration and desalting. Available in several diameters and molecular weight limits.

Concentrates 2 mL down to 15-70 μl98% sample recoveryFast processing time - 10 to 30 minute spin timesEngineered dead stopReverse spin capability provides consistent recoveriesApplications:Concentration of biological samples containing antigens, antibodies, enzymes, nucleic acids, or microorganismsPurification of macromolecular components found in tissue culture extracts or cell lysatesPrimer removal, PCR cleanup, removal of linkers or macromolecular labels from a reaction mix, and protein removal prior to HPLCDesalting, buffer exchange and protein dialysisMaterial Supplied:The Amicon™ Ultra-2 device is supplied with two microcentrifuge tubes. During operation, one tube is used to collect filtrate; the other tube is to recover the concentrated sample.

Specifications

Disposable Single-use
Material (Membrane) High-recovery Ultracel-PL Regenerated Cellulose
MWCO 100 kDa
Sterility Non-sterile
Filtration Area 1 cm2
For Use With (Application) Protein purification, protein concentration and desalting
Housing Material Regenerated Cellulose (RC)
Includes Two microcentrifuge tubes
Quantity 24Pack
Type Centrifugal Filters
Volume (Metric) 2 mL
Length (Metric) 119.7 mm
Diameter (Metric) 15.9 mm
Show More Show Less