EMD Millipore Amicon™ Pro Purification System
Collapses the protein purification workflow into as few as three spins, in a single device
Manufacturer:  EMD Millipore ACS510002
 View more versions of this product
Catalog No. ACS510002
Specifications Description & Specifications
Specifications
Purification System
12cm
2/Pk.
2 Amicon Ultra NMWCO 100KDa filters, 2 Exchange devices, and 2 Collection tubes with cap
100KDa