Please login to your online account to display your discounted pricing
EMD Millipore Amicon™ Pro Purification System
Collapses the protein purification workflow into as few as three spins, in a single device
Manufacturer: emd millipore  ACS505002
 View more versions of this product
Catalog No. ACS505002
  • / Pack of 2 Each
Description & Specifications
Specifications
Type Purification System
Includes 2 Amicon Ultra NMWCO 50KDa filters, 2 Exchange devices, and 2 Collection tubes with cap
Length (Metric) 12cm
MWCO 50KDa
Quantity 2/Pk.