Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Pro Affinity concentration Kit Protein G with Amicon™ Ultra-0.5 Device


Manufacturer: emd millipore  ACK5003PG

 View more versions of this product

Catalog No. ACK5003PG

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Membrane Material Ultracel™ low binding regenerated cellulose
MWCO 3 kd
Length (Metric) 12cm
Diameter (Metric) 29.7mm
Volume (Metric) 10mL
Diameter (English) 1.2 in.
Color Clear with pink cap
Type Amicon™ Pro Affinity Concentration Kit Protein G with 3 kd Amicon™ Ultra-0.5 Device
Includes 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 3 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer
Quantity 12/Pk.
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Length (English) 4.7 in.
Surface Area 1cm2
Housing Material Polypropylene