missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MilliporeSigma™ Amicon™ Pro Affinity concentration Kit Protein G with Amicon™ Ultra-0.5 Device

Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying and concentrating Antibodies.

$477.00 - $477.00

Specifications

Color Clear with pink cap
Diameter (English) 1.2 in.
Diameter (Metric) 29.7mm
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Housing Material Polypropylene
View More Specs

Includes:
  • 12/pk Amicon Pro
  • 12/pk Amicon Ultra
  • 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose)
  • 2 X 66mL Bind/Wash Buffer
  • 12mL Elution buffer
  • 2.5mL Neutralization buffer

Products 5
Catalog Number Mfr. No. Includes MWCO Type Price Quantity  
Catalog Number Mfr. No. Includes MWCO Type Price Quantity  
ACK5003PG View Documents MilliporeSigma™
ACK5003PG
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 3 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer 3 kd Amicon™ Pro Affinity Concentration Kit Protein G with 3 kd Amicon™ Ultra-0.5 Device Pack of 12 Each for $477.00
 
ACK5010PG View Documents MilliporeSigma™
ACK5010PG
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 10 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer 10 kd Amicon™ Pro Affinity Concentration Kit Protein G with 10 kd Amicon™ Ultra-0.5 Device Pack of 12 Each for $477.00
 
ACK5030PG View Documents MilliporeSigma™
ACK5030PG
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer 30 kd Amicon™ Pro Affinity Concentration Kit Protein G with 30 kd Amicon™ Ultra-0.5 Device Pack of 12 Each for $477.00
 
ACK5050PG View Documents MilliporeSigma™
ACK5050PG
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 50 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer 50 kd Amicon™ Pro Affinity Concentration Kit Protein G with 50 kd Amicon™ Ultra-0.5 Device Pack of 12 Each for $477.00
 
ACK5100PG View Documents MilliporeSigma™
ACK5100PG
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 100 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer 100 kd Amicon™ Pro Affinity Concentration Kit Protein G with 100 kd Amicon™ Ultra-0.5 Device Pack of 12 Each for $477.00
 
Description

Description

This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL Protein G resin (Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer

Order Info

Reagents available. (See Cat. No. ACR5000PG)

Specifications

Specifications

Clear with pink cap
29.7mm
Polypropylene
12cm
12/Pk.
10mL
1.2 in.
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
4.7 in.
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
1cm2