MilliporeSigma™ Amicon™ Pro Affinity concentration Kit Protein G with Amicon™ Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Protein G - A Centrifugal Tool for purifying and concentrating Antibodies.
Manufacturer: MilliporeSigma™ ACK5030PG
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL Protein G resin (Binding capacity: 20mg human IgG/ml agarose), 2 X 66ml Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization bufferSpecifications
Polypropylene | |
10mL | |
Clear with pink cap | |
29.7mm | |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer | |
12cm | |
30 kd | |
1cm2 |
Amicon™ Pro Affinity Concentration Kit Protein G with 30 kd Amicon™ Ultra-0.5 Device | |
Ultracel™ low binding regenerated cellulose | |
1.2 in. | |
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment | |
4.7 in. | |
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin | |
12/Pk. |