Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Pro Affinity concentration Kit Protein G with Amicon™ Ultra-0.5 Device


Manufacturer: emd millipore  ACK5010PG

 View more versions of this product

Catalog No. ACK5010PG

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Includes 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 10 kd, 3mL Protein G resin (Binding capacity: 20mg human IgG/mL agarose), 2 x 66mL Bind/Wash Buffer, 12mL Elution buffer, 2.5mL Neutralization buffer
Surface Area 1cm2
Diameter (Metric) 29.7mm
Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Type Amicon™ Pro Affinity Concentration Kit Protein G with 10 kd Amicon™ Ultra-0.5 Device
Length (English) 4.7 in.
MWCO 10 kd
Quantity 12/Pk.
Color Clear with pink cap
Length (Metric) 12cm
Housing Material Polypropylene
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Diameter (English) 1.2 in.
Volume (Metric) 10mL
Membrane Material Ultracel™ low binding regenerated cellulose