MilliporeSigma™ Amicon™ Pro Affinity concentration Kit Ni-NTA with Amicon™ Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Ni-NTA - A Centrifugal Tool for purifying and concentrating His-tagged proteins.
$361.00 - $361.00
Specifications
Membrane Material Ultracel™ low binding regenerated cellulose
Housing Material Polypropylene
Length (Metric) 12cm
Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
View More Specs
Includes:
  • 12/pk Amicon Pro
  • 12/pk Amicon Ultra
  • 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose)
  • 25mL Bind Buffer (4X)
  • 25mL Wash buffer (4X)
  • 5mL elution buffer (4X)
Products 4
Catalog Number Mfr. No. Includes MWCO Price Quantity  
Catalog Number Mfr. No. Includes MWCO Price Quantity  
ACK5003NT View Documents MilliporeSigma™
ACK5003NT
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 3 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25mL Wash buffer (4x), 5mL elution buffer (4x) 3 kd Pack of 12 Each for $361.00
ACK5030NT View Documents MilliporeSigma™
ACK5030NT
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x) 30 kd Pack of 12 Each for $361.00
ACK5050NT View Documents MilliporeSigma™
ACK5050NT
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 100 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x) 50 kd Pack of 12 Each for $361.00
ACK5100NT View Documents MilliporeSigma™
ACK5100NT
12/Pk. Amicon Pro, 12/pk Amicon Ultra NMWCO 50 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x) 100 kd Pack of 12 Each for $361.00
Description
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25ml Bind Buffer (4X), 25mL Wash buffer (4X), 5ml elution buffer (4X)
Order Info
Reagents available. (See Cat. No. ACR5000NT)
Specifications
Specifications
Ultracel™ low binding regenerated cellulose
Polypropylene
12cm
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin