MilliporeSigma™ Amicon™ Pro Affinity concentration Kit Ni-NTA with Amicon™ Ultra-0.5 Device
Amicon Pro Affinity Concentration Kit Ni-NTA - A Centrifugal Tool for purifying and concentrating His-tagged proteins.
Manufacturer:  MilliporeSigma™ ACK5030NT
 View more versions of this product
Catalog No. ACK5030NT
Description
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25ml Bind Buffer (4X), 25mL Wash buffer (4X), 5ml elution buffer (4X)
Specifications
Specifications
Polypropylene
10mL
Clear with pink cap
29.7mm
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x)
12cm
30 kd
1cm2
Amicon™ Pro Affinity Concentration Kit Ni-NTA with 30 kd Amicon™ Ultra-0.5 Device
Ultracel™ low binding regenerated cellulose
1.2 in.
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
4.7 in.
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
12/Pk.