Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Pro Affinity concentration Kit Ni-NTA with Amicon™ Ultra-0.5 Device


Manufacturer: emd millipore  ACK5050NT

 View more versions of this product

Catalog No. ACK5050NT

  • / Pack of 12 Each
 

Description & Specifications

Specifications

For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Length (English) 4.7 in.
Housing Material Polypropylene
Volume (Metric) 10mL
Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Includes 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 100 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x)
Diameter (Metric) 29.7mm
Type Amicon™ Pro Affinity Concentration Kit Ni-NTA with 50 kd Amicon™ Ultra-0.5 Device
Diameter (English) 1.2 in.
Length (Metric) 12cm
MWCO 50 kd
Color Clear with pink cap
Membrane Material Ultracel™ low binding regenerated cellulose
Surface Area 1cm2
Quantity 12/Pk.