Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Pro Affinity concentration Kit Ni-NTA with Amicon™ Ultra-0.5 Device


Manufacturer: emd millipore  ACK5010NT

 View more versions of this product

Catalog No. ACK5010NT

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Max. Force 4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
Length (English) 4.7 in.
Length (Metric) 12cm
Membrane Material Ultracel™ low binding regenerated cellulose
Type Amicon™ Pro Affinity Concentration Kit Ni-NTA with 10 kd Amicon™ Ultra-0.5 Device
Diameter (English) 1.2 in.
Diameter (Metric) 29.7mm
For Use With (Application) Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
Housing Material Polypropylene
Includes 12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 10 kd, 3mL HisBind Ni-NTA resin (Binding capacity: 5 to 10mg His tagged protein/ml agarose), 25mL Bind Buffer (4x), 25 mL Wash buffer (4x), 5mL elution buffer (4x)
MWCO 10 kd
Quantity 12/Pk.
Surface Area 1cm2
Volume (Metric) 10mL
Color Clear with pink cap