MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra-0.5 Device
A Centrifugal Tool for purifying and concentrating proteins.
Manufacturer: MilliporeSigma™ ACK5003GS
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose), 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5mL GST Reconstitution buffer (10X)Specifications
Polypropylene | |
10mL | |
Clear with pink cap | |
29.7mm | |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 3 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | |
12cm | |
3 kd | |
1cm2 |
Amicon™ Pro Affinity Concentration Kit GST with 3 kd Amicon™ Ultra-0.5 Device | |
Ultracel™ low binding regenerated cellulose | |
1.2 in. | |
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment | |
4.7 in. | |
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin | |
12/Pk. |