missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra-0.5 Device
A Centrifugal Tool for purifying and concentrating proteins.
$426.00 - $426.00
Specifications
Diameter (English) | 1.2 in. |
---|---|
Diameter (Metric) | 29.7 mm |
Material (Membrane) | Ultracel Low-binding Regenerated Cellulose |
For Use With (Application) | Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment |
Housing Material | Polypropylene |
Includes:
- 12/pk Amicon Pro
- 12/pk Amicon Ultra
- 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose)
- 125mg Glutathione (reduced, free acid)
- 25mL GST Bind/Wash Buffer (10X)
- 5mL GST Reconstitution buffer (10X)
Catalog Number | Mfr. No. | Includes | MWCO | Type | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | MWCO | Type | Price | Quantity | |||
ACK5003GS
|
MilliporeSigma™
ACK5003GS |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 3 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | 3 kDa | Amicon Pro Affinity Concentration Kit GST with 3 kd Amicon Ultra-0.5 Device |
Pack of 12 Each for $426.00
|
|
ACK5010GS
|
MilliporeSigma™
ACK5010GS |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 10 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | 10 kDa | Amicon Pro Affinity Concentration Kit GST with 10 kd Amicon Ultra-0.5 Device |
Pack of 12 Each for $426.00
|
|
ACK5030GS
|
MilliporeSigma™
ACK5030GS |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 30 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | 30 kDa | Amicon Pro Affinity Concentration Kit GST with 30 kd Amicon Ultra-0.5 Device |
Pack of 12 Each for $426.00
|
|
ACK5050GS
|
MilliporeSigma™
ACK5050GS |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 50 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | 50 kDa | Amicon Pro Affinity Concentration Kit GST with 50 kd Amicon Ultra-0.5 Device |
Pack of 12 Each for $426.00
|
|
ACK5100GS
|
MilliporeSigma™
ACK5100GS |
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 100 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x) | 100 kDa | Amicon Pro Affinity Concentration Kit GST with 100 kd Amicon Ultra-0.5 Device |
Pack of 12 Each for $426.00
|
|
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose), 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5mL GST Reconstitution buffer (10X)Order Info
Reagents available. (See Cat. No. ACR5000NT)
Specifications
1.2 in. | |
Ultracel Low-binding Regenerated Cellulose | |
Polypropylene | |
12 cm |
29.7 mm | |
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment | |
4.7 in. | |
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin |