MilliporeSigma™ Amicon™ Pro Affinity concentration Kit GST with Amicon™ Ultra-0.5 Device
A Centrifugal Tool for purifying and concentrating proteins.
Manufacturer:  MilliporeSigma™ ACK5010GS
 View more versions of this product
Catalog No. ACK5010GS
Description
Description
This kit includes 12/pk Amicon Pro, 12/pk Amicon Ultra NMWCO, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/ml agarose), 125 mg Glutathione (reduced, free acid), 25ml GST Bind/Wash Buffer (10X), 5mL GST Reconstitution buffer (10X)
Specifications
Specifications
Polypropylene
10mL
Clear with pink cap
29.7mm
12/Pk. Amicon Pro, 12/Pk. Amicon Ultra NMWCO 10 kd, 3mL GST Bind resin (Binding capacity: 5 to 8mg GST tagged protein/mL agarose), 125mg Glutathione (reduced, free acid), 25mL GST Bind/Wash Buffer (10x), 5mL GST Reconstitution buffer (10x)
12cm
10 kd
1cm2
Amicon™ Pro Affinity Concentration Kit GST with 10 kd Amicon™ Ultra-0.5 Device
Ultracel™ low binding regenerated cellulose
1.2 in.
Antibody Labeling, Protein Purification, Protein Concentration, Buffer Exchange, Protein Enrichment
4.7 in.
4000xg for concentration and buffer exchange spin; 1000xg for flow through, wash, elution and reverse spin
12/Pk.