Please login to your online account to display your discounted pricing

EMD Millipore Amicon™ Bioseparations Stirred Cells†

Concentrate, desalt, and purify aqueous biological solutions

$1,035.00 - $1,470.00

Specifications

For Use With (Application) Protein purification, protein concentration, protein desalting
View More Specs
Includes:  Connection for 0.25 in. dia. (6.4mm) pressure tubing; max. operating pressure, 75psi (5.3 bar).
Requires:  Magnetic stirrer required for operation.
Products
Catalog Number Mfr. No. Dimensions (L x W x H) Filtration Area Price Quantity    

5123

 
emd millipore
5123
3.5 x 3.5 x 5.03 in. (9 x 9 x 12.8cm) 28.7cm2 Each for $1,470.00

5122

 
emd millipore
5122
2.7 x 2.7 x 3.8 in. (7 x 7 x 9.8cm) 13.4cm2 Each for $1,159.00

5121

 
emd millipore
5121
2.38 x 2.38 x 3 in. (6 x 6 x 7.7cm) 4.1cm2 Each for $1,035.00
Description & Specifications

Specifications

For Use With (Application) Protein purification, protein concentration, protein desalting

Process from 3mL up to 400mL of biological solutions using either concentration or diafiltration operating modes.

  • Sturdy construction; autoclavable Consist of clear pressurized vessel with integral magnetic stirring bar, nylon cap and tube fitting assembly, polysulfone body and membrane holder, acetal/polysulfone stirrer, silicone O-rings, and black aluminum stand
Concentration Mode
  • Gas pressure is applied directly to ultrafiltration cell Solutes above membrane molecular weight cutoff are retained in cell, while water and solutes below cutoff pass into filtrate
Diafiltration Mode
    Ultrafiltration cell is connected to auxiliary reservoir containing diafiltrate solution; reservoir is then gas pressurized Fluid volume and macrosolute concentration remain constant as filtrate is replaced by diafiltrate solution Provides a simple means for rapid microsolute exchange
  • Typically used as a substitute for dialysis

Applications:

Perform concentration, desalting, recovery or exclusion; Recover monoclonal antibodies and growth factors from tissue culture supernatants; Concentration and buffer exchange of chromatography fractions

Use with Millipore Ultrafiltration Membranes.