missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore Albumin-Glycerol Fixative

Used for mounting tissue sections

Manufacturer:  EMD Millipore R0320374

 View more versions of this product

Catalog No. MR0320374


 

Specifications Description & Specifications

Specifications

Poly Bottle
290°C
Clear to Milky
Glycerin 50%, Egg Albumin 49%, Sodium Salicylate 1%
17.9°C
Mounting tissue sections
Albumin Glycerol Fixative
500mL