missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Aervent-50 Disposable Filters
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
Manufacturer: MilliporeSigma™ MTGR85000
Description
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
- 50mm PTFE membrane
- Hydrophobic
- High flowrates and throughput, low extractables, low particle shedding
- Broad chemical compatibility
Applications:
Sterile venting of vessels and carboys, autoclave vacuum break lines, low volume sterile filtration of non-aqueous fluids, small fermenter inlet and exhaust gas filtration
Specifications
Autoclavable | |
Polypropylene | |
Both sides: hose connection for 6.5 to 9.5mm I.D. silicone tubing | |
100/Pk. | |
62 mm |
Yes | |
0.2 μm | |
Both sides: hose connection for 6.5 to 9.5mm I.D. silicone tubing | |
Syringe Filter |