MilliporeSigma™ Aervent-50 Disposable Filters
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
Manufacturer: MilliporeSigma™ MTGR75000
Description
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
- 50mm PTFE membrane
- Hydrophobic
- High flowrates and throughput, low extractables, low particle shedding
- Broad chemical compatibility
Applications:
Sterile venting of vessels and carboys, autoclave vacuum break lines, low volume sterile filtration of non-aqueous fluids, small fermenter inlet and exhaust gas filtration
Specifications
Yes | |
62 mm | |
Hydrophobic PTFE | |
100/Pk. |
Syringeless Filter | |
3mm (1/8 in.) NPTM | |
0.2 μm |