missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Aervent-50 Disposable Filters
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
Manufacturer: MilliporeSigma™ MTGR05000
Description
Sterilizing-grade filters designed for critical small-scale gas line and air venting applications
- 50mm PTFE membrane
- Hydrophobic
- High flowrates and throughput, low extractables, low particle shedding
- Broad chemical compatibility
Applications:
Sterile venting of vessels and carboys, autoclave vacuum break lines, low volume sterile filtration of non-aqueous fluids, small fermenter inlet and exhaust gas filtration
Specifications
Yes | |
62 mm | |
Both sides: hose connection for 4 to 9.5mm I.D. latex tubing and female Luer slip | |
Both sides: hose connection for 4 to 9.5mm I.D. latex tubing and female Luer slip | |
100/Pk. |
Syringe Filter | |
Yes | |
Polypropylene | |
0.2 μm |