Please login to your online account to display your discounted pricing
EMD Millipore Accessories for Pellicon XL Devices
Manufacturer: emd millipore  XXMULTIMN
 View more versions of this product
Catalog No. XXMULTIMN
  • / Each
Description & Specifications
Specifications
Product Type Multi-manifold Kit
Quantity 1 Pack
For Use With (Equipment) Pellicon™ XL 50 cassette and Labscale™ TFF system
For Use With (Application) Mounts directly onto either Labscale™ system reservoir or standard peristaltic pump to allow simultaneous use of three Pellicon™ XL devices