EMD Millipore 47mm Glass Vacuum Filter Holders
Designed to support bacteriological analysis and analysis of particulates in water and other fluids. EMD Millipore 47mm Glass Vacuum Filter Holders include a glass funnel and base, aluminum clamp and silicone stopper. Available with PTFE-coated sealing surfaces or a stainless steel filter support.
$254.00 - $363.00
Specifications
Capacity (Metric) 300mL
Diameter (Metric) Inner
  • 47mm Filter
  • 76mm Funnel Base
Surface Area (Metric) 9.6cm2
View More Specs
Alert:
Not recommended for use with flammable liquids
Products ${productFamilyLength}
Catalog Number Mfr. No. Autoclavable Material Price Quantity  
Catalog Number Mfr. No. Autoclavable Material Price Quantity  
XX1004700 Accessories EMD Millipore
XX1004700
Yes - without filter
  • Borosilicate glass funnel and base
  • Polytetrafluoroethylene seals
  • Anodized aluminum spring clamp
  • Silicone stopper
Each for $254.00
XX1004720 Accessories EMD Millipore
XX1004720
Yes - with filter
  • Polytetrafluoroethylene (PTFE)-faced, borosilicate glass funnel and base
  • Polytetrafluoroethylene seals
  • Anodized aluminum spring clamp
  • Silicone stopper
Each for $363.00
XX1004730 Accessories EMD Millipore
XX1004730
Yes - without filter
  • Borosilicate glass funnel and base
  • Polytetrafluoroethylene seals
  • Stainless steel screen
  • Anodized aluminum spring clamp
  • Silicone stopper
Each for $363.00
Specifications
Description
  • Consists of borosilicate glass base and 300mL funnel, 47mm filter support, anodized aluminum spring clamp and silicone stopper
  • No. 8 perforated silicone stopper mounts in standard 1 L and 4 L filtering flasks
  • Sanitize with sulfuric acid or hydrochloric acid; sterilize with UV sterilizer (2 to 3 min.) or autoclave (20 min. at 121°C)
Glass Filter Holder
  • Ideal for bacteriological analysis of water (with membrane filters) and for analysis of suspended solids in water (using depth filters)
Glass Filter Holder with PTFE-Coated Sealing Surfaces
  • Useful for bacteriological analysis applications where filter holder must be autoclaved with membrane in place
  • Coated sealing surfaces prevent membrane adhesion and filter damage during autoclaving
  • Features polytetrafluoroethylene (PTFE)-faced funnel and base
Glass Filter Holder with Stainless-Steel Support
  • For use in particulate contamination analysis of oils and hydraulic fluids by gravimetric or particulate counting methods
  • Also suitable for use in exfoliative cytology applications
  • Includes stainless-steel screen filter support
Description & Specifications
Specifications
300mL
  • 47mm Filter
  • 76mm Funnel Base
9.6cm2