MilliporeSigma™ Aerosol Filter Holders, 47 mm
Vacuum-filter air or other gases from clean-room/open atmospheres/from closed systems for particulate contamination analysis.
Supplier: MilliporeSigma™ XX5004710
Description
These aerosol filter holders are designed for monitoring airborne particulates from the environment and/or closed systems. Choose the holder design most appropriate for your method. Aerosol filter holders are available in two formats: open-type for open atmosphere sampling and a standard design with a closed connection and inlet dispersion chamber. Applications:Particulate Contamination Monitoring in GasesSpecifications
Stainless Steel Bodies, Filter Support Screens; Anodized Aluminum Locking Rings; Nylon Lockwheels | |
Vacuum-Filter Air or Other Gases From Open Atmospheres for Particulate Contamination Analysis | |
4 in. | |
2.7 in. | |
6.9 cm | |
1.1 cm |
9.6 cm2 | |
Laboratory Filter Holders or Cyclones | |
10.2 cm | |
0.43 in. | |
6.9 cm |