Please login to your online account to display your discounted pricing

EMD Millipore 25mm Glass Microanalysis Vacuum Filter Holder and Support

Vacuum filter small volumes for particulate or biological contamination analysis

$314.00 - $347.00

Specifications

Capacity (Metric) 15mL
Diameter (Metric) Inner 25mm
Surface Area (Metric) 2.5cm2
View More Specs
Includes:  Borosilicate glass funnel and base; fritted glass or removable stainless-steel filter support; anodized aluminum spring clamp; No. 5 stopper
Products
Catalog Number Mfr. No. Material Price Quantity    

XX1002500

  Accessories
emd millipore
XX1002500
Fritted Glass Each for $314.00

XX1002530

  Accessories
emd millipore
XX1002530
Stainless Steel Support Each for $347.00
Description & Specifications

Specifications

Capacity (Metric) 15mL
Diameter (Metric) Inner 25mm
Surface Area (Metric) 2.5cm2

  • Funnel capacity: 15mL
  • Filtration area: 2.5cm2
  • Not autoclavable with filter in place