missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore 25mm Glass Microanalysis Vacuum Filter Holder and Support

Vacuum filter small volumes for particulate or biological contamination analysis

Manufacturer:  EMD Millipore XX1002530

 View more versions of this product

Catalog No. XX1002530


 

Specifications

Description

Support, Filter; EMD Millipore; With stainless-steel support; For glass microanalysis vacuum filter holder and support; Vacuum filter small volumes for particulate or biological contamination analysis; 25mm

  • Funnel capacity: 15mL
  • Filtration area: 2.5cm2
  • Not autoclavable with filter in place

Description & Specifications

Specifications

Stainless Steel Support
25mm
15mL
2.5cm2
Accessories