Please login to your online account to display your discounted pricing

EMD Millipore 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts


Manufacturer: emd millipore  XX1002514

 View more versions of this product

Catalog No. XX1002514

  • / Each
 

Description & Specifications

Specifications

Capacity (Metric) 15mL
Material Glass
Description Funnel, 15mL
For Use With (Equipment) 25mm Glass Microanalysis Vacuum Filter