missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts

Accessories for filter holders for sample preparation.

Manufacturer:  EMD Millipore XX1002505

 View more versions of this product

Catalog No. XX1002505


 

Specifications

Description

Flask, Filtering; EMD Millipore; Made of glass; For 25mm glass microanalysis vacuum filter accessories and replacement parts; Capacity: 125mL

  • Fitting Outlet: No. 5 perforated silicone stopper mounts
  • Diameter: 25mm
  • Filtration Area: 2.5 sq. cm
Description & Specifications

Specifications

Filtering flask, 125mL
Glass
25mm Glass Microanalysis Vacuum Filter
125mL