Please login to your online account to display your discounted pricing
EMD Millipore 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts
Manufacturer: emd millipore  XX5002501
 View more versions of this product
Catalog No. XX5002501
  • / Each
Description & Specifications
Specifications
Description Support Screen (for XX10 025 30)
Material Stainless Steel
For Use With (Equipment) 25mm Glass Microanalysis Vacuum Filter
  • Fitting Outlet: No. 5 perforated silicone stopper mounts
  • Diameter: 25mm
  • Filtration Area: 2.5 sq. cm