EMD Millipore 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts
Accessories for filter holders for sample preparation.
Manufacturer:  EMD Millipore XX1002542
 View more versions of this product
Catalog No. XX1002542
Specifications
Description
Base; EMD Millipore; With screen, gasket and stopper; For 25mm glass microanalysis vacuum filter accessories and replacement parts
  • Fitting Outlet: No. 5 perforated silicone stopper mounts
  • Diameter: 25mm
  • Filtration Area: 2.5 sq. cm
Description & Specifications
Specifications
Base, stopper, screen and gasket
Stainless Steel (screen)
25mm Glass Microanalysis Vacuum Filter