Please login to your online account to display your discounted pricing
EMD Millipore 25mm Glass Microanalysis Vacuum Filter Accessories and Replacement Parts
Manufacturer: emd millipore  XX1002535
 View more versions of this product
Catalog No. XX1002535
  • / Pack of 10 Each
Description & Specifications
Specifications
Description Support Screen Gasket
Material PTFE
For Use With (Equipment) 25mm Glass Microanalysis Vacuum Filter
  • Fitting Outlet: No. 5 perforated silicone stopper mounts
  • Diameter: 25mm
  • Filtration Area: 2.5 sq. cm