missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Nylon Membrane

Overmolded polypropylene housing and optimized housing design

$283.00 - $1,884.00

Specifications

Color Code Purple
Applications For filtering aqueous and organic solutions
Diameter (Metric) 13mm
Type Syringe Filter
Material Nylon (NYL)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Outlet Style Pore Size Price Quantity  
Catalog Number Mfr. No. Outlet Style Pore Size Price Quantity  
SLGNX13NL EMD Millipore
SLGNX13NL
male Luer slip 0.2μm Pack of 100 Each for $283.00
SLGNX13NK EMD Millipore
SLGNX13NK
male Luer slip 0.2μm Pack of 1000 Each for $1,884.00
SLHNX13NL EMD Millipore
SLHNX13NL
male Luer slip 0.45μm Pack of 100 Each for $283.00
SLHNX13NK EMD Millipore
SLHNX13NK
male Luer slip 0.45μm Pack of 1000 Each for $1,853.00
SLHNX13TL EMD Millipore
SLHNX13TL
Tube outlet 0.45μm Pack of 100 Each for $287.00
Specifications

Description

  • Nylon membranes provide broad chemical compatibility for use with aqueous and organic solutions
  • Overmolded polypropylene housing design provides a more robust filter, with a maximum housing pressure of 150psig (10 bar) enabling solutions to be filtered faster
  • Optimized housing design maintains 50% less hold volume than traditional 13mm filters to ensure maximum sample recovery
  • Fully automated manufacturing process minimizes the risk of contamination and meets pharmaceutical standards
  • Color-coded band on housing clearly indicates membrane type

Order Info

Inlet/outlet connections: female Luer-Lok™/male Luer slip.

Description & Specifications

Specifications

Purple
13mm
Nylon (NYL)
For filtering aqueous and organic solutions
Syringe Filter