missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EMD Millipore 13mm Nonsterile Millex™ Syringe Filters - Hydrophobic PTFE Membrane

Overmolded polypropylene housing and optimized housing design

$293.00 - $2,008.00

Specifications

Applications For Preparation of organic solutions
Diameter (Metric) 13mm
Housing Material Polypropylene
Material Hydrophobic Polytetrafluoroethylene (PTFE)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Outlet Style Pore Size Quantity Type Price Quantity  
Catalog Number Mfr. No. Outlet Style Pore Size Quantity Type Price Quantity  
SLFGX13NL EMD Millipore
SLFGX13NL
male Luer slip 0.2μm 100/Pk. Syringe Filter FG Pack of 100 Each for $293.00
SLFHX13NK EMD Millipore
SLFHX13NK
male Luer slip 0.45μm 1000/Pk. Syringe Filter FH Pack of 1000 Each for $1,915.00
SLFHX13NL EMD Millipore
SLFHX13NL
Male Luer slip 0.45μm 100/Pk. Syringe Filter FH Pack of 100 Each for $293.00
SLFGX13NK EMD Millipore
SLFGX13NK
male Luer slip 0.22μm 1000/Pk. Syringe Filter FG Pack of 1000 Each for $2,008.00
SLFGX13TL EMD Millipore
SLFGX13TL
Male Tube Outlet 0.2μm 100/Pk. Syringe Filter FG Pack of 100 Each for $316.00
Specifications

Description

  • Hydrophobic PTFE membranes have excellent solvent resistance and are ideal for HPLC sample preparation
  • Overmolded polypropylene housing design provides a more robust filter, with a maximum housing pressure of 150psig (10 bar) enabling solutions to be filtered faster
  • Optimized housing design maintains 50% less hold volume than traditional 13mm filters to ensure maximum sample recovery
  • Fully automated manufacturing process minimizes the risk of contamination and meets pharmaceutical standards
  • Color-coded band on housing clearly indicates membrane type

Order Info

Inlet/outlet connections: female Luer-Lok™/male Luer slip.

Description & Specifications

Specifications

For Preparation of organic solutions
13mm
Polypropylene
Hydrophobic Polytetrafluoroethylene (PTFE)